Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAGTGTACTGCAGTTTTACCCGAA[A/G]GCTAATATCGTTGCCTACCATGACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615661 MIM: 613295 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PDCD2L PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PDCD2L - programmed cell death 2 like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
UBA2 - ubiquitin like modifier activating enzyme 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005499.2 | 328 | Silent Mutation | AAA,AAG | K,K 86 | NP_005490.1 | |
XM_005258404.3 | 328 | Silent Mutation | AAA,AAG | K,K 198 | XP_005258461.2 | |
XM_006722962.2 | 328 | Silent Mutation | AAA,AAG | K,K 58 | XP_006723025.1 | |
XM_011526304.2 | 328 | Silent Mutation | AAA,AAG | K,K 198 | XP_011524606.1 | |
XM_017026134.1 | 328 | Silent Mutation | AAA,AAG | K,K 198 | XP_016881623.1 |