Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATTGGTTGAATCCATGGATACAGA[A/C]CCCATGCACTGTATACTCTGTCTGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIR643 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MIR643 - microRNA 643 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF766 - zinc finger protein 766 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001010851.2 | Intron | NP_001010851.1 | ||||
XM_011527460.1 | Intron | XP_011525762.1 | ||||
XM_017027440.1 | Intron | XP_016882929.1 | ||||
XM_017027441.1 | Intron | XP_016882930.1 |