Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGTCATCACGGAGCAGCCCAAGCA[A/G]CGCGGCATGCGCTTCCGCTACGAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604758 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RELB PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RELB - RELB proto-oncogene, NF-kB subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006509.3 | 536 | Silent Mutation | CAA,CAG | Q,Q 135 | NP_006500.2 | |
XM_005259127.3 | 536 | Silent Mutation | CAA,CAG | Q,Q 132 | XP_005259184.1 | |
XM_005259128.2 | 536 | Silent Mutation | CAA,CAG | Q,Q 135 | XP_005259185.1 |