Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGATCAACGCTGCATTCTCATCCTT[C/G]AGGCGCTGGTTGTCAGCGCGGAGGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614987 MIM: 613245 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
EPS8L1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EPS8L1 - EPS8 like 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PPP1R12C - protein phosphatase 1 regulatory subunit 12C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271618.1 | 2344 | Silent Mutation | CTC,CTG | L,L 764 | NP_001258547.1 | |
NM_017607.3 | 2344 | Silent Mutation | CTC,CTG | L,L 766 | NP_060077.1 | |
XM_005259013.3 | 2344 | Silent Mutation | CTC,CTG | L,L 765 | XP_005259070.1 | |
XM_011527045.2 | 2344 | Silent Mutation | CTC,CTG | L,L 765 | XP_011525347.1 |