Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAGGAGGAGTGGAAATGCCTGGAC[C/G]CTGGGCAGAAAGCTTTATACAGGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF534 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF534 - zinc finger protein 534 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001143938.1 | 140 | Missense Mutation | CCT,GCT | P,A 27 | NP_001137410.1 | |
NM_001143939.1 | 140 | Missense Mutation | CCT,GCT | P,A 27 | NP_001137411.1 | |
NM_001291368.1 | 140 | Missense Mutation | CCT,GCT | P,A 27 | NP_001278297.1 | |
NM_001291369.1 | 140 | Missense Mutation | CCT,GCT | P,A 27 | NP_001278298.1 |