Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACACAACTTATAAAATGTTCATGGC[C/T]GCAGCAGGAGTAGAGGGTGAGGTTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
PRR19 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PRR19 - proline rich 19 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM145 - transmembrane protein 145 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_173633.2 | 724 | Silent Mutation | GCC,GCT | A,A 210 | NP_775904.2 | |
XM_005258781.4 | 724 | Silent Mutation | GCC,GCT | A,A 224 | XP_005258838.1 | |
XM_011526791.2 | 724 | Silent Mutation | GCC,GCT | A,A 210 | XP_011525093.1 | |
XM_011526792.1 | 724 | Silent Mutation | GCC,GCT | A,A 224 | XP_011525094.1 |