Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGGTGCCATTTTGTGTATCTTCTG[C/T]ACCTAAGGAGGAAAAAAGATAGAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611168 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DUXA PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DUXA - double homeobox A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001012729.1 | 592 | Missense Mutation | ACA,GCA | T,A 183 | NP_001012747.1 |
ZIM3 - zinc finger imprinted 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |