Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGGGTGCCTATGACCTGAGGCGGC[A/G]GGAGAGGCAGTCCCGCCAGACGTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 162815 MIM: 610391 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AZU1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AZU1 - azurocidin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001700.4 | 287 | Missense Mutation | CAG,CGG | Q,R 89 | NP_001691.1 |
PLPPR3 - phospholipid phosphatase related 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |