Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCGTGCACCTTACCACGTAGTTGAG[A/G]CTCTCGGCCACCGAATGGTCGCGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600696 MIM: 602518 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ECH1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ECH1 - enoyl-CoA hydratase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001398.2 | 955 | Silent Mutation | AGC,AGT | S,S 290 | NP_001389.2 | |
XM_017026448.1 | 955 | Silent Mutation | AGC,AGT | S,S 210 | XP_016881937.1 |
LGALS4 - galectin 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |