Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGGGGTGGAGGTGGGAGATCAAAT[A/G]TGCATTCAGAGATGACCATGGTGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600493 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADGRE1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ADGRE1 - adhesion G protein-coupled receptor E1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256252.1 | Intron | NP_001243181.1 | ||||
NM_001256253.1 | Intron | NP_001243182.1 | ||||
NM_001256254.1 | Intron | NP_001243183.1 | ||||
NM_001256255.1 | Intron | NP_001243184.1 | ||||
NM_001974.4 | Intron | NP_001965.3 | ||||
XM_011527794.1 | Intron | XP_011526096.1 |