Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGGAGAGACCCTGACCATAGAGAC[A/G]GGGTGGCCGGGCCTGGGGCTGGGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610481 MIM: 614715 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SHD PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SHD - Src homology 2 domain containing transforming protein D | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMIGD2 - transmembrane and immunoglobulin domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001169126.1 | 451 | Silent Mutation | CCC,CCT | P,P 251 | NP_001162597.1 | |
NM_001308232.1 | 451 | Silent Mutation | CCC,CCT | P,P 135 | NP_001295161.1 | |
NM_144615.2 | 451 | Silent Mutation | CCC,CCT | P,P 255 | NP_653216.2 | |
XM_017026284.1 | 451 | Intron | XP_016881773.1 |