Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGCACATTGTGCTTCTGCAAGGCT[C/T]GCCGGACCACGCTGGGGGCTTTGTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616743 | ||||||||||||||||||||
Literature Links: |
RGL3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RGL3 - ral guanine nucleotide dissociation stimulator like 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001035223.3 | 2019 | Missense Mutation | CAA,CGA | Q,R 646 | NP_001030300.3 | |
NM_001161616.2 | 2019 | Missense Mutation | CAA,CGA | Q,R 652 | NP_001155088.2 |