Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTAAGTAAATAAAATCTATTGTAG[A/G]CTTTCCCACATTCGTTACATATATA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 194628 | ||||||||||||||||||||
Literature Links: |
ZNF121 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZNF121 - zinc finger protein 121 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001008727.3 | 1359 | Missense Mutation | GCC,GTC | A,V 376 | NP_001008727.1 | |
NM_001308269.1 | 1359 | Missense Mutation | GCC,GTC | A,V 376 | NP_001295198.1 | |
XM_017027239.1 | 1359 | Missense Mutation | GCC,GTC | A,V 376 | XP_016882728.1 |