Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCCCCCACCAGGCCTCACACTCCC[C/T]GACTTGCCCGAGCAGTTCTCCCCAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612258 MIM: 603157 | ||||||||||||||||||||
Literature Links: |
MAST3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MAST3 - microtubule associated serine/threonine kinase 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PIK3R2 - phosphoinositide-3-kinase regulatory subunit 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005027.3 | 876 | Silent Mutation | CCC,CCT | P,P 112 | NP_005018.1 |