Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGCAGGAGCGCGGTTCTGACTGC[A/G]CCAGTCAAAAGTGACCAGCGCGCCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603832 MIM: 606862 | ||||||||||||||||||||
Literature Links: |
NDUFA3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NDUFA3 - NADH:ubiquinone oxidoreductase subunit A3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
OSCAR - osteoclast associated, immunoglobulin-like receptor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282349.1 | 958 | UTR 3 | NP_001269278.1 | |||
NM_001282350.1 | 958 | UTR 3 | NP_001269279.1 | |||
NM_130771.4 | 958 | Missense Mutation | CGC,TGC | R,C 256 | NP_570127.3 | |
NM_133168.4 | 958 | Missense Mutation | CGC,TGC | R,C 241 | NP_573398.2 | |
NM_133169.4 | 958 | Missense Mutation | CGC,TGC | R,C 252 | NP_573399.2 | |
NM_206818.2 | 958 | UTR 3 | NP_996554.2 |