Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCCTGTGATTCCCTTGGATGGATC[G/T]GTGAAAATCCAGTGCCAGGCCATTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 147045 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FCAR PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FCAR - Fc fragment of IgA receptor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002000.3 | 293 | Silent Mutation | TCG,TCT | S,S 44 | NP_001991.1 | |
NM_133269.3 | 293 | Silent Mutation | TCG,TCT | S,S 44 | NP_579803.1 | |
NM_133271.3 | 293 | Silent Mutation | TCG,TCT | S,S 44 | NP_579805.1 | |
NM_133272.3 | 293 | Silent Mutation | TCG,TCT | S,S 32 | NP_579806.1 | |
NM_133273.3 | 293 | Silent Mutation | TCG,TCT | S,S 32 | NP_579807.1 | |
NM_133274.3 | 293 | Silent Mutation | TCG,TCT | S,S 32 | NP_579808.1 | |
NM_133277.3 | 293 | Intron | NP_579811.1 | |||
NM_133278.3 | 293 | Silent Mutation | TCG,TCT | S,S 32 | NP_579812.1 | |
XM_011526625.2 | 293 | Silent Mutation | TCG,TCT | S,S 17 | XP_011524927.1 | |
XM_017026473.1 | 293 | Silent Mutation | TCG,TCT | S,S 17 | XP_016881962.1 | |
XM_017026474.1 | 293 | Silent Mutation | TCG,TCT | S,S 17 | XP_016881963.1 |