Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCACACTGTCCTCCACTCGGTAGCG[C/G]ATCTGGTATTTGGCTTGAAAGAGGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604237 | ||||||||||||||||||||
Literature Links: |
C19orf60 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C19orf60 - chromosome 19 open reading frame 60 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CRLF1 - cytokine receptor like factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004750.4 | 1090 | Missense Mutation | ATC,ATG | I,M 274 | NP_004741.1 | |
XM_011528422.2 | 1090 | Missense Mutation | ATC,ATG | I,M 252 | XP_011526724.1 | |
XM_011528423.2 | 1090 | Missense Mutation | ATC,ATG | I,M 274 | XP_011526725.1 | |
XM_011528424.2 | 1090 | Missense Mutation | ATC,ATG | I,M 252 | XP_011526726.1 |