Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATGCTCAGTGAGATATGCTTTGCC[A/G]TTAAAGGCTTTGCCACATTCCTTAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613749 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF260 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF260 - zinc finger protein 260 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001012756.2 | 1354 | Silent Mutation | AAC,AAT | N,N 146 | NP_001012774.1 | |
NM_001166036.1 | 1354 | Silent Mutation | AAC,AAT | N,N 146 | NP_001159508.1 | |
NM_001166037.1 | 1354 | Silent Mutation | AAC,AAT | N,N 146 | NP_001159509.1 | |
NM_001166038.1 | 1354 | Silent Mutation | AAC,AAT | N,N 146 | NP_001159510.1 | |
XM_011526897.2 | 1354 | Silent Mutation | AAC,AAT | N,N 146 | XP_011525199.1 | |
XM_017026741.1 | 1354 | Silent Mutation | AAC,AAT | N,N 146 | XP_016882230.1 |