Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCCTCTGGACAAGCCGGGCTGGCC[A/G]GCCCCCACAGCTGGTCCTGGACCTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606651 MIM: 611011 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GRIN3B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GRIN3B - glutamate ionotropic receptor NMDA type subunit 3B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_138690.2 | 609 | Missense Mutation | CAG,CGG | Q,R 197 | NP_619635.1 | |
XM_017026243.1 | 609 | Intron | XP_016881732.1 |
LOC105372235 - uncharacterized LOC105372235 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM259 - transmembrane protein 259 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
WDR18 - WD repeat domain 18 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |