Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGGGTCTTCATCAGTTTGAACGTGC[C/T]GGTGACCTCCATGGCGTCCTGCAGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603314 | ||||||||||||||||||||
Literature Links: |
SLC27A5 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SLC27A5 - solute carrier family 27 member 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321196.1 | 2025 | Missense Mutation | AGC,GGC | S,G 555 | NP_001308125.1 | |
NM_012254.2 | 2025 | Missense Mutation | AGC,GGC | S,G 639 | NP_036386.1 | |
XM_011526364.1 | 2025 | Missense Mutation | CAG,CGG | Q,R 595 | XP_011524666.1 | |
XM_017026214.1 | 2025 | Intron | XP_016881703.1 |
ZNF446 - zinc finger protein 446 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |