Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGGTTCCGACAGTCAAGATGGCGG[A/G]AGCAGCTACCCAGGCTTCCCTGGAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603182 MIM: 609615 | ||||||||||||||||||||
Literature Links: |
ILF3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ILF3 - interleukin enhancer binding factor 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
QTRT1 - queuine tRNA-ribosyltransferase catalytic subunit 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_031209.2 | 33 | Missense Mutation | GAA,GGA | E,G 3 | NP_112486.1 |