Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTTGACCAGGAATCTGGACAGGTC[C/T]TGCAGGATGGGCTCGCTGTGCAAGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602781 MIM: 601778 | ||||||||||||||||||||
Literature Links: |
HCN2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HCN2 - hyperpolarization activated cyclic nucleotide gated potassium channel 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001194.3 | Intron | NP_001185.3 |
POLRMT - RNA polymerase mitochondrial | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005035.3 | Intron | NP_005026.3 | ||||
XM_005259580.3 | Intron | XP_005259637.2 |