Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCGTGTGTCTCCGCAGCATCCGTC[G/T]GTGTCCAGGGCCAGTCTGTGTCAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANKRD27 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ANKRD27 - ankyrin repeat domain 27 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032139.2 | 3202 | Silent Mutation | AGA,CGA | R,R 1016 | NP_115515.2 | |
XM_006723412.2 | 3202 | Silent Mutation | AGA,CGA | R,R 1016 | XP_006723475.1 | |
XM_017027349.1 | 3202 | Silent Mutation | AGA,CGA | R,R 820 | XP_016882838.1 |