Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATTTGAAGCTCTGTATACATTCAA[A/G]AGATTTTTCTCTCATGTGTACTTCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616841 | ||||||||||||||||||||
Literature Links: |
ZNF468 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZNF468 - zinc finger protein 468 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001008801.1 | 1009 | Intron | NP_001008801.1 | |||
NM_001277120.1 | 1009 | Intron | NP_001264049.1 | |||
XM_017027443.1 | 1009 | Missense Mutation | XP_016882932.1 | |||
XM_017027444.1 | 1009 | Missense Mutation | XP_016882933.1 |