Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCACACACACGGCAGCACCGGCAC[A/C]GCGGTGAGCCTGCAGGGTGGGCTGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 615240 | ||||||||||||||||||||
Literature Links: |
KCTD15 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KCTD15 - potassium channel tetramerization domain containing 15 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001129994.1 | 365 | Silent Mutation | ACA,ACC | T,T 21 | NP_001123466.1 | |
NM_001129995.1 | 365 | Silent Mutation | ACA,ACC | T,T 21 | NP_001123467.1 | |
NM_024076.2 | 365 | Silent Mutation | ACA,ACC | T,T 21 | NP_076981.2 | |
XM_011527296.1 | 365 | UTR 5 | XP_011525598.1 | |||
XM_011527297.2 | 365 | Silent Mutation | ACA,ACC | T,T 21 | XP_011525599.1 | |
XM_011527298.2 | 365 | Silent Mutation | ACA,ACC | T,T 21 | XP_011525600.1 | |
XM_017027283.1 | 365 | Silent Mutation | ACA,ACC | T,T 21 | XP_016882772.1 | |
XM_017027284.1 | 365 | Intron | XP_016882773.1 |