Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACCTGGGGGCGGGTGGTCTTGGGA[C/G]CTGCCTGCCGAGCATTCATGCTGGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607157 | ||||||||||||||||||||
Literature Links: |
SIGLEC11 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SIGLEC11 - sialic acid binding Ig like lectin 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135163.1 | 1844 | Intron | NP_001128635.1 | |||
NM_052884.2 | 1844 | Intron | NP_443116.2 | |||
XM_005258476.3 | 1844 | Silent Mutation | AGC,AGG | S,R 604 | XP_005258533.2 | |
XM_005258477.3 | 1844 | Intron | XP_005258534.2 |