Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCGCATGCCATCGCGCACCCGCAC[A/G]CAGAAATCTGTGCTGTAGCAGTCGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606048 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MBOAT7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MBOAT7 - membrane bound O-acyltransferase domain containing 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001146056.2 | 1272 | Silent Mutation | TGC,TGT | C,C 237 | NP_001139528.1 | |
NM_001146082.2 | 1272 | Silent Mutation | TGC,TGT | C,C 310 | NP_001139554.1 | |
NM_001146083.2 | 1272 | Silent Mutation | TGC,TGT | C,C 237 | NP_001139555.1 | |
NM_024298.4 | 1272 | Silent Mutation | TGC,TGT | C,C 310 | NP_077274.3 | |
XM_011527299.2 | 1272 | Silent Mutation | TGC,TGT | C,C 310 | XP_011525601.1 | |
XM_011527300.2 | 1272 | Silent Mutation | TGC,TGT | C,C 310 | XP_011525602.1 | |
XM_017027296.1 | 1272 | Silent Mutation | TGC,TGT | C,C 310 | XP_016882785.1 |
TMC4 - transmembrane channel like 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |