Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGCAATTAAGGAATGAACAACCCA[C/T]GATTTCTCCACACACACTGCTTTTA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ZNF442 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZNF442 - zinc finger protein 442 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_030824.2 | 412 | Missense Mutation | ATG,GTG | M,V 125 | NP_110451.1 | |
XM_006722908.3 | 412 | Missense Mutation | ATG,GTG | M,V 118 | XP_006722971.1 | |
XM_017027316.1 | 412 | Missense Mutation | ATG,GTG | M,V 133 | XP_016882805.1 | |
XM_017027317.1 | 412 | Missense Mutation | ATG,GTG | M,V 68 | XP_016882806.1 | |
XM_017027318.1 | 412 | Missense Mutation | ATG,GTG | M,V 125 | XP_016882807.1 |
ZNF563 - zinc finger protein 563 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |