Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGAAGCCACGTTCCCATCTTCCAC[A/G]GCAGAATATTCCAGGAGAGCTTCAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604955 MIM: 604947 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KIR2DS4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KIR2DS4 - killer cell immunoglobulin like receptor, two Ig domains and short cytoplasmic tail 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
KIR3DL2 - killer cell immunoglobulin like receptor, three Ig domains and long cytoplasmic tail 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001242867.1 | 374 | Intron | NP_001229796.1 | |||
NM_006737.3 | 374 | Intron | NP_006728.2 | |||
XM_011526940.2 | 374 | Missense Mutation | AGC,GGC | S,G 73 | XP_011525242.1 | |
XM_017026784.1 | 374 | Missense Mutation | AGC,GGC | S,G 73 | XP_016882273.1 |