Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCTGGTGCACCCCCAGTGGGTGCT[C/G]ACAGCTGCCCATTGCCTAAAGAAGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 147960 | ||||||||||||||||||||
Literature Links: |
KLK2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KLK2 - kallikrein related peptidase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001002231.2 | 224 | Silent Mutation | CTC,CTG | L,L 61 | NP_001002231.1 | |
NM_001256080.1 | 224 | Intron | NP_001243009.1 | |||
NM_005551.4 | 224 | Silent Mutation | CTC,CTG | L,L 61 | NP_005542.1 |
KLKP1 - kallikrein pseudogene 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |