Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCTCCACTATGAATTCTGTGATGG[A/C]TTGCAAGGTTTGAACTCTGACTTTA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ZNF347 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZNF347 - zinc finger protein 347 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001172674.1 | 2932 | Missense Mutation | AAG,AAT | K,N 835 | NP_001166145.1 | |
NM_001172675.1 | 2932 | Missense Mutation | AAG,AAT | K,N 835 | NP_001166146.1 | |
NM_032584.2 | 2932 | Missense Mutation | AAG,AAT | K,N 834 | NP_115973.2 | |
XM_005259335.4 | 2932 | Missense Mutation | AAG,AAT | K,N 834 | XP_005259392.1 | |
XM_005259336.4 | 2932 | Missense Mutation | AAG,AAT | K,N 776 | XP_005259393.1 | |
XM_017027384.1 | 2932 | Missense Mutation | AAG,AAT | K,N 834 | XP_016882873.1 | |
XM_017027385.1 | 2932 | Missense Mutation | AAG,AAT | K,N 776 | XP_016882874.1 | |
XM_017027386.1 | 2932 | Missense Mutation | AAG,AAT | K,N 776 | XP_016882875.1 |
ZNF415 - zinc finger protein 415 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |