Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGTGTGGGTCGGCAGGATGCGGAA[C/G]TGCAGCAGGGTGAGCGCCAGGACCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611517 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CYP4F11 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CYP4F11 - cytochrome P450 family 4 subfamily F member 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001128932.1 | 1922 | Missense Mutation | CAC,CAG | H,Q 488 | NP_001122404.1 | |
NM_021187.3 | 1922 | Missense Mutation | CAC,CAG | H,Q 488 | NP_067010.3 |