Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACGCAGCGCACCATCAACAGCACCA[A/G]CACCACGAGCAGGCAGAGCACGAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600135 MIM: 605076 MIM: 605058 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CTXN1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CTXN1 - cortexin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_206833.3 | 344 | Silent Mutation | CTG,TTG | L,L 42 | NP_996664.1 |
SNAPC2 - small nuclear RNA activating complex polypeptide 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TGFBR3L - transforming growth factor beta receptor 3 like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TIMM44 - translocase of inner mitochondrial membrane 44 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |