Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCACATGATACAGCCACTGGGCTTA[C/T]ACAGTATGCATTGGACCAGGGCGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600538 MIM: 606034 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PRDX2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PRDX2 - peroxiredoxin 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RNASEH2A - ribonuclease H2 subunit A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006397.2 | 619 | Missense Mutation | ACA,ATA | T,I 125 | NP_006388.2 | |
XM_006722619.2 | 619 | Missense Mutation | ACA,ATA | T,I 81 | XP_006722682.1 |