Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGGCCCTAAGTCCTCTGGTTGTC[A/G]AGGGGTAAGTCCCCAGGGTCTGGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600128 MIM: 179490 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC102725254 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC102725254 - uncharacterized LOC102725254 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PDE4C - phosphodiesterase 4C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_011528056.2 | 2091 | Silent Mutation | CTC,CTT | L,L 601 | XP_011526358.1 | |
XM_011528058.2 | 2091 | Silent Mutation | CTC,CTT | L,L 476 | XP_011526360.1 | |
XM_017026866.1 | 2091 | Silent Mutation | CTC,CTT | L,L 707 | XP_016882355.1 |
RAB3A - RAB3A, member RAS oncogene family | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |