Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATGGGGAACGATTCTGTCAGCTAC[A/G]AGTATGGGGATTACAGCGACCTCTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609949 MIM: 615475 | ||||||||||||||||||||
Literature Links: |
C5AR2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C5AR2 - complement component 5a receptor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271749.1 | 166 | Missense Mutation | AAG,GAG | K,E 9 | NP_001258678.1 | |
NM_001271750.1 | 166 | Missense Mutation | AAG,GAG | K,E 9 | NP_001258679.1 | |
NM_018485.2 | 166 | Missense Mutation | AAG,GAG | K,E 9 | NP_060955.1 | |
XM_011526736.2 | 166 | Missense Mutation | AAG,GAG | K,E 9 | XP_011525038.1 |
DHX34 - DEAH-box helicase 34 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |