Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTGCGAGAACGACGACCTGGCCAC[C/G]AGCCTCGTCCTGGACCCCTACCTCG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 613198 | ||||||||||||||||||||
Literature Links: |
COX6B2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
COX6B2 - cytochrome c oxidase subunit 6B2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
KMT5C - lysine methyltransferase 5C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032701.3 | 305 | Silent Mutation | ACC,ACG | T,T 19 | NP_116090.2 | |
XM_005259338.3 | 305 | UTR 5 | XP_005259395.1 | |||
XM_006723442.3 | 305 | Intron | XP_006723505.1 | |||
XM_011527415.2 | 305 | Silent Mutation | ACC,ACG | T,T 19 | XP_011525717.1 | |
XM_017027388.1 | 305 | UTR 5 | XP_016882877.1 |
TMEM150B - transmembrane protein 150B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |