Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCAATATCTCAAGCTGATCACTGG[A/G]TTCTGAGGCCCAGAAGGGAGACTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616802 MIM: 616804 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TARM1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TARM1 - T cell-interacting, activating receptor on myeloid cells 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135686.2 | 653 | Missense Mutation | NP_001129158.2 | |||
XM_005278278.3 | 653 | Intron | XP_005278335.1 |
VSTM1 - V-set and transmembrane domain containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |