Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGCTTACGGTCGCAGAAGGCGTCG[C/T]TCCAGCGACCGGAGCCCCGCATCAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 151445 MIM: 609565 | ||||||||||||||||||||
Literature Links: |
FCER2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FCER2 - Fc fragment of IgE receptor II | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001207019.2 | 887 | Missense Mutation | AAC,AGC | N,S 268 | NP_001193948.2 | |
NM_001220500.1 | 887 | Missense Mutation | AAC,AGC | N,S 269 | NP_001207429.1 | |
NM_002002.4 | 887 | Missense Mutation | AAC,AGC | N,S 269 | NP_001993.2 | |
XM_005272462.4 | 887 | Missense Mutation | AAC,AGC | N,S 269 | XP_005272519.1 |
MCEMP1 - mast cell expressed membrane protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TRAPPC5 - trafficking protein particle complex 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |