Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATAGATTTTTCCCAGGAAGAATGGC[A/T]ATGGATGAACCCTGCTCAGAAGCGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ZNF471 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZNF471 - zinc finger protein 471 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321768.1 | 219 | Intron | NP_001308697.1 | |||
NM_020813.3 | 219 | Missense Mutation | CAA,CTA | Q,L 29 | NP_065864.2 | |
XM_011527148.1 | 219 | Missense Mutation | CAA,CTA | Q,L 29 | XP_011525450.1 | |
XM_017027032.1 | 219 | Intron | XP_016882521.1 |