Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCTTCCCTTCCCTTTTGGGCTCAG[G/T]TTACCCAGTTTTTGCCCCCTCTCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612082 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CIC PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CIC - capicua transcriptional repressor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304815.1 | Intron | NP_001291744.1 | ||||
NM_015125.4 | Intron | NP_055940.3 | ||||
XM_005258673.2 | Intron | XP_005258730.1 | ||||
XM_005258674.2 | Intron | XP_005258731.1 | ||||
XM_005258675.2 | Intron | XP_005258732.1 | ||||
XM_011526660.2 | Intron | XP_011524962.1 | ||||
XM_011526661.2 | Intron | XP_011524963.1 | ||||
XM_011526662.2 | Intron | XP_011524964.1 | ||||
XM_011526663.2 | Intron | XP_011524965.1 | ||||
XM_011526664.2 | Intron | XP_011524966.1 | ||||
XM_011526665.2 | Intron | XP_011524967.1 | ||||
XM_011526666.2 | Intron | XP_011524968.1 | ||||
XM_011526667.2 | Intron | XP_011524969.1 |