Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGGCTTATCTTTATCCCTTACACT[C/G]AAGGCTTTCTAACCAGTATGAATTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614275 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF565 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF565 - zinc finger protein 565 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001042474.2 | 2314 | UTR 3 | NP_001035939.1 | |||
NM_152477.4 | 2314 | UTR 3 | NP_689690.3 | |||
XM_011526512.2 | 2314 | UTR 3 | XP_011524814.1 | |||
XM_011526514.2 | 2314 | UTR 3 | XP_011524816.1 | |||
XM_017026341.1 | 2314 | UTR 3 | XP_016881830.1 | |||
XM_017026342.1 | 2314 | UTR 3 | XP_016881831.1 | |||
XM_017026343.1 | 2314 | UTR 3 | XP_016881832.1 |