Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGTGCCAGCCGCGTCCCTCGTACA[C/G]GTAGCCGTCCGAGCCCACCACGAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608199 MIM: 616561 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PGLYRP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PGLYRP2 - peptidoglycan recognition protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_052890.3 | 1502 | Missense Mutation | CTG,GTG | L,V 457 | NP_443122.3 | |
XM_006722633.3 | 1502 | Missense Mutation | CTG,GTG | L,V 457 | XP_006722696.1 |
RASAL3 - RAS protein activator like 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |