Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAACACAAAATTTTAAGATGTCCA[C/T]CTGGGAGGACACGGACTTCAAAAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604086 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF155 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF155 - zinc finger protein 155 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF221 - zinc finger protein 221 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001297588.1 | Intron | NP_001284517.1 | ||||
NM_001297589.1 | Intron | NP_001284518.1 | ||||
NM_013359.2 | Intron | NP_037491.2 | ||||
XM_017027232.1 | Intron | XP_016882721.1 | ||||
XM_017027233.1 | Intron | XP_016882722.1 |