Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGGAGAACCAGGAGCACCAGCTGG[C/G]GAACAACACCCTCTCCAGCCACCTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605418 MIM: 601729 | ||||||||||||||||||||
Literature Links: |
DKKL1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DKKL1 - dickkopf like acrosomal protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001197301.1 | 354 | Intron | NP_001184230.1 | |||
NM_001197302.1 | 354 | Intron | NP_001184231.1 | |||
NM_014419.3 | 354 | Missense Mutation | GCG,GGG | A,G 96 | NP_055234.1 | |
XM_006723142.2 | 354 | Missense Mutation | GCG,GGG | A,G 111 | XP_006723205.1 | |
XM_011526724.1 | 354 | Missense Mutation | GCG,GGG | A,G 119 | XP_011525026.1 | |
XM_011526725.1 | 354 | Missense Mutation | GCG,GGG | A,G 103 | XP_011525027.1 | |
XM_011526726.2 | 354 | Missense Mutation | GCG,GGG | A,G 103 | XP_011525028.1 | |
XM_011526727.2 | 354 | Missense Mutation | GCG,GGG | A,G 119 | XP_011525029.1 | |
XM_011526728.2 | 354 | Intron | XP_011525030.1 | |||
XM_011526729.1 | 354 | Missense Mutation | GCG,GGG | A,G 21 | XP_011525031.1 | |
XM_017026593.1 | 354 | Missense Mutation | GCG,GGG | A,G 21 | XP_016882082.1 |
LOC101928295 - uncharacterized LOC101928295 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TEAD2 - TEA domain transcription factor 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |