Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCAGAGAACAACTCCACACCCTCA[C/T]GGGAAGGGAAGAGACAGACACGCTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601155 MIM: 180664 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARHGAP45 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ARHGAP45 - Rho GTPase activating protein 45 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001258328.2 | 1281 | Intron | NP_001245257.1 | |||
NM_001282334.1 | 1281 | Intron | NP_001269263.1 | |||
NM_001282335.1 | 1281 | Intron | NP_001269264.1 | |||
NM_001321232.1 | 1281 | Intron | NP_001308161.1 | |||
NM_012292.4 | 1281 | Intron | NP_036424.2 | |||
XM_006722713.1 | 1281 | Intron | XP_006722776.1 | |||
XM_011527858.1 | 1281 | Intron | XP_011526160.1 | |||
XM_011527860.2 | 1281 | Intron | XP_011526162.1 |
POLR2E - RNA polymerase II subunit E | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001316323.1 | 1281 | UTR 3 | NP_001303252.1 | |||
NM_001316324.1 | 1281 | UTR 3 | NP_001303253.1 | |||
NM_002695.3 | 1281 | UTR 3 | NP_002686.2 | |||
XM_011528070.2 | 1281 | UTR 3 | XP_011526372.1 | |||
XM_017026883.1 | 1281 | UTR 3 | XP_016882372.1 |