Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTTGAACTGCTTCATTCAGGACTC[C/T]GCAAATTCCCTGAAGTAGAAGGAAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604086 | ||||||||||||||||||||
Literature Links: |
ZNF155 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZNF155 - zinc finger protein 155 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF221 - zinc finger protein 221 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001297588.1 | 230 | Missense Mutation | CGC,TGC | R,C 14 | NP_001284517.1 | |
NM_001297589.1 | 230 | Missense Mutation | CGC,TGC | R,C 14 | NP_001284518.1 | |
NM_013359.2 | 230 | Missense Mutation | CGC,TGC | R,C 14 | NP_037491.2 | |
XM_017027232.1 | 230 | Intron | XP_016882721.1 | |||
XM_017027233.1 | 230 | Intron | XP_016882722.1 |