Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTTTGCTTCTGTTGGAGATGTTCC[C/T]GTCGGATGCTGAAGAACAAGCCTGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 188070 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CACTIN PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CACTIN - cactin, spliceosome C complex subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080543.1 | 3364 | UTR 3 | NP_001074012.1 | |||
NM_021231.1 | 3364 | UTR 3 | NP_067054.1 | |||
XM_011528160.2 | 3364 | Intron | XP_011526462.1 | |||
XM_011528161.2 | 3364 | Intron | XP_011526463.1 |
CACTIN-AS1 - CACTIN antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TBXA2R - thromboxane A2 receptor | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |