Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGATGGGCTCCACCTTGATCTGTGG[C/T]GGCGGGGGCGGTGGGGCTAGCGCCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611888 MIM: 606784 | ||||||||||||||||||||
Literature Links: |
ERF PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ERF - ETS2 repressor factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001301035.1 | 1291 | Silent Mutation | CCA,CCG | P,P 347 | NP_001287964.1 | |
NM_001308402.1 | 1291 | Silent Mutation | CCA,CCG | P,P 347 | NP_001295331.1 | |
NM_001312656.1 | 1291 | Silent Mutation | CCA,CCG | P,P 347 | NP_001299585.1 | |
NM_006494.3 | 1291 | Silent Mutation | CCA,CCG | P,P 422 | NP_006485.2 | |
XM_017026468.1 | 1291 | Silent Mutation | CCA,CCG | P,P 347 | XP_016881957.1 | |
XM_017026469.1 | 1291 | Silent Mutation | CCA,CCG | P,P 347 | XP_016881958.1 |
GSK3A - glycogen synthase kinase 3 alpha | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |