Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGATGGGGTCTCCACAGATTCCTT[C/G]CATCCCAAATGGAGGGTGCCCTCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605546 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GP6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GP6 - glycoprotein VI platelet | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001083899.2 | 1268 | Missense Mutation | GCA,GGA | A,G 415 | NP_001077368.2 | |
NM_001256017.2 | 1268 | UTR 3 | NP_001242946.2 | |||
NM_016363.5 | 1268 | UTR 3 | NP_057447.5 |